Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037806.3 | NP_001032895.2 | 3 - 4 | 325 | 63 | |
| XM_005269050.3 | XP_005269107.1 | 3 - 4 | 650 | 63 | ||
| XM_006719525.2 | XP_006719588.1 | 3 - 4 | 620 | 63 | ||
| XM_011538610.2 | XP_011536912.1 | 4 - 5 | 404 | 63 | ||
| XM_011538611.2 | XP_011536913.1 | 4 - 5 | 374 | 63 | ||
| GenBank mRNA | AK093900.1 | - | 1 - 2 | 1798 | 63 | |
| BC110599.2 | - | 1 - 2 | 1469 | 63 | ||
| CN359946.1 | - | 3 - 4 | 325 | 63 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000335999.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000433948.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000477361.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000480927.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | ENST00000491441.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010640.hg.1 | NM_001037806.3 | Refseq |

NCKAP5L
Genomic Map