| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012249.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012249.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17012249.hg.1 |
TC17000931.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000417245.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009179.hg.1 |
NM_012418.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
ENST00000334850.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
ENST00000527221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
NM_001077182.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700169556.hg.1 |
TC1700009179.hg.1 |
Clariom D TC |