Assay Details
| Gene Symbol: MUC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002457.3 | NP_002448.3 | 13290 | 56 | ||
| GenBank mRNA | AF257167.1 | - | 3 - 4 | 711 | 56 | |
| L21998.1 | - | 13290 | 56 | |||
| M94132.1 | - | 3 - 4 | 678 | 56 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000616479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | ENST00000616479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000361558.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000613187.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000616479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | ENST00000616479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006506.hg.1 | NM_002457.4 | Refseq |

MUC2
Genomic Map