Assay Details
| Gene Symbol: GLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006708.2 | NP_006699.2 | 5 - 6 | 586 | 127 | |
| GenBank mRNA | AB209801.1 | - | 5 - 6 | 477 | 127 | |
| AK129573.1 | - | 5 - 6 | 519 | 127 | ||
| AK130312.1 | - | 5 - 6 | 483 | 127 | ||
| AK293345.1 | - | 5 - 6 | 506 | 127 | ||
| AK312662.1 | - | 5 - 6 | 555 | 127 | ||
| AL833442.1 | - | 5 - 6 | 536 | 127 | ||
| BC001741.1 | - | 5 - 6 | 527 | 3352922 | 127 | |
| BC011365.1 | - | 5 - 6 | 540 | 4182593 | 127 | |
| BC015934.1 | - | 5 - 6 | 511 | 3912185 | 127 | |
| BT019987.1 | - | 5 - 6 | 464 | 127 | ||
| BT019988.1 | - | 5 - 6 | 464 | 127 | ||
| D13315.1 | - | 5 - 6 | 551 | 127 | ||
| FJ224325.1 | - | 5 - 6 | 551 | 127 | ||
| KJ534846.1 | - | 5 - 6 | 464 | 127 | ||
| L07837.1 | - | 5 - 6 | 524 | 127 | ||
| S83285.1 | - | 5 - 6 | 473 | 127 | ||
More Information
Set Membership: |
|

GLO1
Genomic Map