Assay Details
| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018942.2 | NP_061815.2 | 1 - 2 | 599 | 84 | |
| GenBank mRNA | M99587.1 | - | 1 - 2 | 589 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018942.2 | NP_061815.2 | 1 - 2 | 599 | 84 | |
| GenBank mRNA | M99587.1 | - | 1 - 2 | 589 | 84 | |
Set Membership: |
|