Assay Details
| Gene Symbol: LINC01138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097058.1 | - | 1 - 2 | 208 | 84 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501290 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501291 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20534446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501280 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000381475.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000417554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000417554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000417554.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440199.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440862.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440862.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440862.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000440862.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000442423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000442423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000442423.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000613452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000613452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000613452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000613452.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000614292.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | ENST00000622328.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_027468.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018475.hg.1 | NR_104014.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009816.hg.1 | ENST00000415381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009816.hg.1 | ENST00000415381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009816.hg.1 | ENST00000415381.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009816.hg.1 | ENST00000415381.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007530 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050883 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051505 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007530 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A050883 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051505 | - | - |

LINC01138
Genomic Map