Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010857.hg.1 |
ENST00000328449.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010857.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010857.hg.1 |
TC16000744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010856.hg.1 |
ENST00000328449.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010856.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010856.hg.1 |
TC16000744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010860.hg.1 |
ENST00000328449.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010860.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010860.hg.1 |
TC16000744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010859.hg.1 |
ENST00000328449.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010859.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16010859.hg.1 |
TC16000744.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168155.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168155.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168155.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168155.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168155.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009008.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168148.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168149.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168149.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168149.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168149.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168149.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168151.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
ENST00000621771.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168153.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600168154.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168154.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168154.hg.1 |
ENST00000328449.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600168154.hg.1 |
NM_207419.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600168154.hg.1 |
TC1600009008.hg.1 |
Clariom D TC |