Assay Details
| Gene Symbol: YPEL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145524.1 | NP_001138996.1 | 4 - 5 | 452 | 95 | |
| NM_031477.4 | NP_113665.3 | 3 - 4 | 1098 | 95 | ||
| GenBank mRNA | AB098737.1 | - | 4 - 5 | 452 | 95 | |
| AF305622.1 | - | 4 - 5 | 455 | 95 | ||
| BC005009.1 | - | 4 - 5 | 408 | 3639517 | 95 | |
| BC050664.1 | - | 3 - 4 | 1097 | 5748830 | 95 | |
More Information
Set Membership: |
|

YPEL3
Genomic Map