Assay Details
Gene Symbol: TAF15-NR4A3 fusion | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF162670.1 | - | 149 | 110 | ||
AJ243810.1 | - | 107 | 110 | |||
AJ245932.1 | - | 100 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
