Assay Details
| Gene Symbol: MLL-GAS7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF231995.1 | - | 269 | 150 | ||
| AF231996.1 | - | 361 | 150 | |||
| AF231997.1 | - | 361 | 150 | |||
| AF231998.1 | - | 475 | 264 | |||
| AF231999.1 | - | 475 | 264 | |||
More Information
Set Membership: |
|

MLL-GAS7 fusion
Genomic Map