Assay Details
| Gene Symbol: MLL-GAS7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF231998.1 | - | 405 | 130 | ||
| AF231999.1 | - | 405 | 130 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-GAS7 fusion| Gene Symbol: MLL-GAS7 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF231998.1 | - | 405 | 130 | ||
| AF231999.1 | - | 405 | 130 | |||
Set Membership: |
|