Assay Details
| Gene Symbol: MLL-GMPS fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF297747.1 | - | 401 | 108 | ||
| AF297749.1 | - | 260 | 108 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-GMPS fusion| Gene Symbol: MLL-GMPS fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF297747.1 | - | 401 | 108 | ||
| AF297749.1 | - | 260 | 108 | |||
Set Membership: |
|