Assay Details
| Gene Symbol: MLL-MAML2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ972402.1 | - | 617 | 126 | ||
| DQ084494.1 | - | 165 | 126 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-MAML2 fusion| Gene Symbol: MLL-MAML2 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AJ972402.1 | - | 617 | 126 | ||
| DQ084494.1 | - | 165 | 126 | |||
Set Membership: |
|