Assay Details
| Gene Symbol: ACYP2-ZNF585B fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098695.1 | - | 191 | 84 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACYP2-ZNF585B fusion| Gene Symbol: ACYP2-ZNF585B fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098695.1 | - | 191 | 84 | ||