Assay Details
| Gene Symbol: RALA-OR2W3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL080170.1 | - | 255 | 121 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000005257.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000434466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000436179.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000466491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | ENST00000468201.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007271.hg.1 | NM_005402.3 | Refseq |

RALA-OR2W3 fusion
Genomic Map