Assay Details
| Gene Symbol: MLL-KIAA0826 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ299938.1 | - | 261 | 224 | ||
| DQ299939.1 | - | 147 | 110 | |||
| DQ299940.1 | - | 147 | 110 | |||
More Information
Set Membership: |
|

MLL-KIAA0826 fusion
Genomic Map