Assay Details
| Gene Symbol: PAX3-FOXO1A fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC008826.1 | - | 1386 | 98 | ||
| U02308.1 | - | 761 | 98 | |||
| U02368.1 | - | 1250 | 98 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

PAX3-FOXO1A fusion
Genomic Map