Assay Details
| Gene Symbol: NPM1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D45915.1 | - | 1 | 469 | 122 | |
| U04946.1 | - | 1 | 380 | 122 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPM1-ALK fusion| Gene Symbol: NPM1-ALK fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | D45915.1 | - | 1 | 469 | 122 | |
| U04946.1 | - | 1 | 380 | 122 | ||
Set Membership: |
|