Assay Details
| Gene Symbol: DUX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012146.1 | NP_036278.1 | 1 | 630 | 146 | |
| GenBank mRNA | AJ001481.1 | - | 1 | 630 | 146 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P158257 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 208176_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742311_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742311_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208176_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208201_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208582_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208176_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208176_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208201_at | - | - |
| Human Genome U133A 2.0 Array | 208176_at | - | - |
| Human Genome U133A 2.0 Array | 208201_PM_at | - | - |
| Human Genome U133A 2.0 Array | 208582_PM_s_at | - | - |
| Human Genome U133A 2.0 Array | 208582_s_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_012146_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw538144 | - | - |
| Human Genome U95 Set | 31346_PM_at | - | - |
| Human Genome U95 Set | 31387_at | - | - |
| Human Genome U95 Set | 31387_PM_at | - | - |
| Human Genome U95 Set | 31346_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208176_at4 | - | - |
| Human Genome U133 Set | 208176_PM_at | - | - |
| Human Genome U133 Set | 208176_at | - | - |
| Human Genome U133 Set | 208582_s_at | - | - |
| Human Genome U133 Set | 208201_PM_at | - | - |
| Human Genome U133 Set | 208201_at | - | - |
| Human Genome U133 Set | 208582_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 285085 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 541942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120735-S | NM_012146.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_11120735-S | AJ001481.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_14075-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0005910131 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0005910131 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1990255 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_14075 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1711055 | AJ001481.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1711055 | NM_012146.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1711055 | AJ001481.1 | human_transcriptome |

DUX1
Genomic Map