Assay Details
| Gene Symbol: PABPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080487.2 | NP_001073956.2 | 2 - 3 | 445 | 108 | |
| NM_001294328.1 | NP_001281257.1 | 2 - 3 | 445 | 108 | ||
| XM_017023230.1 | XP_016878719.1 | 4 - 5 | 1700 | 108 | ||
| GenBank mRNA | BC130001.1 | - | 2 - 3 | 445 | 40133435 | 108 |
| BC130002.1 | - | 2 - 3 | 445 | 108 | ||
| EU190484.1 | - | 2 - 3 | 403 | 108 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001346.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16018653.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018653.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018653.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018653.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018653.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | ENST00000427766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018655.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018659.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018659.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018659.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | ENST00000427766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018654.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | ENST00000427766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018656.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187549.hg.1 | TC1600011237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187550.hg.1 | TC1600011237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187554.hg.1 | TC1600011237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187548.hg.1 | TC1600011237.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187551.hg.1 | TC1600011237.hg.1 | Clariom D TC |

PABPN1L
Genomic Map