Assay Details
| Gene Symbol: PABPN1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080487.2 | NP_001073956.2 | 4 - 5 | 620 | 124 | |
| XM_017023230.1 | XP_016878719.1 | 6 - 7 | 1875 | 124 | ||
| GenBank mRNA | EU190484.1 | - | 4 - 5 | 578 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001346.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | ENST00000411789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | ENST00000427766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018651.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018652.hg.1 | ENST00000378358.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018652.hg.1 | ENST00000419291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018652.hg.1 | ENST00000427766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018652.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16018652.hg.1 | TC16001346.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000411789.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011237.hg.1 | NM_001294328.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | ENST00000419291.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | ENST00000547152.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | NM_001080487.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600187547.hg.1 | TC1600011237.hg.1 | Clariom D TC |

PABPN1L
Genomic Map