Assay Details
| Gene Symbol: MLL-AFF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF024541.1 | - | 1 | 91 | 110 | |
| L22179.1 | - | 4374 | 242 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-AFF1 fusion| Gene Symbol: MLL-AFF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF024541.1 | - | 1 | 91 | 110 | |
| L22179.1 | - | 4374 | 242 | |||
Set Membership: |
|