Assay Details
| Gene Symbol: MLL-AFF1 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF024541.1 | - | 2 | 128 | 139 | |
| AF177237.1 | - | 1 | 83 | 94 | ||
| L22179.1 | - | 4411 | 271 | |||
More Information
Set Membership: |
|

MLL-AFF1 fusion
Genomic Map