Assay Details
| Gene Symbol: PTGER2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000956.3 | NP_000947.2 | 1 - 2 | 1089 | 97 | |
| GenBank mRNA | AY275471.1 | - | 1 - 2 | 838 | 97 | |
| BC093927.1 | - | 1 - 2 | 891 | 7939772 | 97 | |
| BC093929.1 | - | 1 - 2 | 891 | 7939774 | 97 | |
| U19487.1 | - | 1 - 2 | 994 | 97 | ||
| X83868.1 | - | 1 - 2 | 838 | 97 | ||
More Information
Set Membership: |
|

PTGER2
Genomic Map