Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309434.1 | NP_001296363.1 | 2 - 3 | 169 | 93 | |
| NM_003134.5 | NP_003125.3 | 2 - 3 | 169 | 93 | ||
| GenBank mRNA | AB061546.1 | - | 2 - 3 | 156 | 93 | |
| AB451391.1 | - | 2 - 3 | 83 | 93 | ||
| BC035495.2 | - | 2 - 3 | 125 | 4298087 | 93 | |
| BC071716.1 | - | 2 - 3 | 105 | 4476669 | 93 | |
| BC100031.1 | - | 2 - 3 | 109 | 2847600 | 93 | |
| DA338824.1 | - | 2 - 3 | 169 | 93 | ||
| U07857.1 | - | 2 - 3 | 104 | 93 | ||
| X73459.1 | - | 2 - 3 | 102 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001228.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001309434.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | ENST00000267884.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | ENST00000558527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | ENST00000560773.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011024.hg.1 | TC15001228.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100008781.hg.1 | ENST00000533701.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100016391.hg.1 | ENST00000599419.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008217.hg.1 | ENST00000513669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165683.hg.1 | TC1500009079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100008781.hg.1 | ENST00000533701.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008217.hg.1 | ENST00000513669.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500165679.hg.1 | TC1500009079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000267884.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558243.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000558720.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559081.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559439.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000559475.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | ENST00000560773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_001309434.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009079.hg.1 | NM_003134.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100016391.hg.1 | ENST00000599419.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 227557 | - | - |

SRP14
Genomic Map