| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07031447.hg.1 |
TC07002047.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07031447.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07031447.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194699.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194699.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194699.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194699.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194699.hg.1 |
TC0700013119.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700194700.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194700.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194700.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194700.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194700.hg.1 |
TC0700013119.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
ENST00000495707.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013119.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194701.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194701.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194701.hg.1 |
ENST00000359321.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700194701.hg.1 |
NM_005431.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700194701.hg.1 |
TC0700013119.hg.1 |
Clariom D TC |