Assay Details
| Gene Symbol: IFNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024013.2 | NP_076918.1 | 1 | 733 | 146 | |
| GenBank mRNA | AB578886.1 | - | 1 | 746 | 146 | |
| V00537.1 | - | 1 | 735 | 146 | ||
| V00538.1 | - | 1 | 733 | 146 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000103.hg.1
Set Membership: |
|

IFNA1
Genomic Map