| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022817.hg.1 |
TC05001680.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022817.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022817.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000383058.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200006586.hg.1 |
ENST00000450724.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022818.hg.1 |
TC05001680.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022818.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05022818.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181118.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181118.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181118.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181118.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181118.hg.1 |
TC0500011730.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC2200006586.hg.1 |
ENST00000450724.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011730.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181117.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181117.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181117.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181117.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181117.hg.1 |
TC0500011730.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000383058.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000416046.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008344.hg.1 |
ENST00000510381.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181116.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181116.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181116.hg.1 |
ENST00000390666.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500181116.hg.1 |
NM_032028.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500181116.hg.1 |
TC0500011730.hg.1 |
Clariom D TC |