Assay Details
| Gene Symbol: C5orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173667.3 | NP_775938.1 | 2 - 3 | 137 | 69 | |
| GenBank mRNA | AK094862.1 | - | 2 - 3 | 109 | 69 | |
| BC111982.1 | - | 1 - 2 | 63 | 8327450 | 69 | |
| BC112008.1 | - | 1 - 2 | 63 | 8327476 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000264.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05003193.hg.1 | TC05000264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003193.hg.1 | ENST00000313303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003193.hg.1 | ENST00000505642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003193.hg.1 | ENST00000510414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003193.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003191.hg.1 | TC05000264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003191.hg.1 | ENST00000505642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003191.hg.1 | ENST00000510414.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003191.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003195.hg.1 | TC05000264.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003195.hg.1 | ENST00000313303.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003195.hg.1 | ENST00000505642.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05003195.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193040.hg.1 | TC0500013172.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193037.hg.1 | TC0500013172.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193038.hg.1 | TC0500013172.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500193043.hg.1 | TC0500013172.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000313303.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000502395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000505642.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000510414.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | ENST00000511407.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NM_173667.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126523.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126524.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126525.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500013172.hg.1 | NR_126525.1 | Refseq |

C5orf64
Genomic Map