Assay Details
Gene Symbol: RPL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000997.4 | NP_000988.1 | 2 - 3 | 244 | 110 | |
GenBank mRNA | AK311827.1 | - | 2 - 3 | 201 | 110 | |
BC079477.1 | - | 1 | 167 | 2819896 | 110 | |
BC084576.1 | - | 1 - 2 | 145 | 110 | ||
BU954721.1 | - | 2 - 3 | 248 | 110 | ||
BX647123.1 | - | 2 - 3 | 171 | 110 | ||
CR456890.1 | - | 1 - 2 | 144 | 110 | ||
D23661.1 | - | 2 - 3 | 172 | 110 | ||
L11567.1 | - | 1 - 2 | 150 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05001300.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
rRNA binding
metal ion binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
rRNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05018339.hg.1 | TC05001300.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018339.hg.1 | ENST00000274242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018339.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007342.hg.1 | ENST00000426602.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018332.hg.1 | TC05001300.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018332.hg.1 | ENST00000274242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018332.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018332.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018332.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05018341.hg.1 | TC05001300.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018341.hg.1 | ENST00000274242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018341.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018341.hg.1 | ENST00000509877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018341.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05018342.hg.1 | TC05001300.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018342.hg.1 | ENST00000274242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018342.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018342.hg.1 | ENST00000509877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018342.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400009349.hg.1 | ENST00000492636.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007704.hg.1 | ENST00000460682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007704.hg.1 | ENST00000464216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007704.hg.1 | ENST00000464216.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012044.hg.1 | ENST00000418887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012044.hg.1 | ENST00000418887.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008208.hg.1 | ENST00000412884.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007222.hg.1 | ENST00000432310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007672.hg.1 | ENST00000456825.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100011351.hg.1 | ENST00000472498.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007319.hg.1 | ENST00000481627.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018015.hg.1 | ENST00000595491.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007580.hg.1 | ENST00000479840.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | TC05001300.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | ENST00000274242.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | ENST00000509877.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05018333.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012244.hg.1 | ENST00000404509.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008687.hg.1 | ENST00000492641.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008179.hg.1 | ENST00000603088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700008179.hg.1 | ENST00000603088.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700012525.hg.1 | ENST00000449336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012044.hg.1 | ENST00000418887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012044.hg.1 | ENST00000418887.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | ENST00000511787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500010585.hg.1 | NR_037665.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400009349.hg.1 | ENST00000492636.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008179.hg.1 | ENST00000603088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700008179.hg.1 | ENST00000603088.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007222.hg.1 | ENST00000432310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173875.hg.1 | TC0500010585.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007319.hg.1 | ENST00000481627.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007580.hg.1 | ENST00000479840.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018015.hg.1 | ENST00000595491.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008208.hg.1 | ENST00000412884.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173885.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173885.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173885.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173885.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173885.hg.1 | TC0500010585.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0700012525.hg.1 | ENST00000449336.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007704.hg.1 | ENST00000460682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007704.hg.1 | ENST00000464216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007704.hg.1 | ENST00000464216.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008687.hg.1 | ENST00000492641.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000274242.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000504562.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000507642.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000508493.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | ENST00000509877.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | NM_000997.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500173877.hg.1 | TC0500010585.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007342.hg.1 | ENST00000426602.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100011351.hg.1 | ENST00000472498.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012244.hg.1 | ENST00000404509.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007672.hg.1 | ENST00000456825.1 | Ensembl |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 116254 | - | - |
