| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157403.hg.1 |
TC0500008384.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500157402.hg.1 |
TC0500008384.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000316788.11 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000395548.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000505423.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000506430.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000507436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000509055.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000514118.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
ENST00000515066.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500008384.hg.1 |
NM_001284.3 |
Refseq |