Assay Details
| Gene Symbol: GPRC5D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018654.1 | NP_061124.1 | 1 - 2 | 899 | 100 | |
| XM_005253421.3 | XP_005253478.1 | 3 - 4 | 3339 | 100 | ||
| XM_017019582.1 | XP_016875071.1 | 2 - 3 | 1432 | 100 | ||
| GenBank mRNA | AF209923.1 | - | 1 - 2 | 899 | 100 | |
| BC069341.1 | - | 1 - 2 | 899 | 7262221 | 100 | |
| BC107077.1 | - | 1 - 2 | 899 | 40027061 | 100 | |
| BC107078.1 | - | 1 - 2 | 899 | 40027066 | 100 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001258.hg.1
Set Membership: |
|

GPRC5D
Genomic Map