Assay Details
| Gene Symbol: OR2M7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004691.1 | NP_001004691.1 | 1 | 879 | 142 | |
| GenBank mRNA | BC136944.1 | - | 1 | 908 | 9020936 | 142 |
| BC136946.1 | - | 1 | 908 | 9020938 | 142 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

OR2M7
Genomic Map