Assay Details
| Gene Symbol: CSH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020991.3 | NP_066271.1 | 5 | 755 | 128 | |
| NM_022644.3 | NP_072170.1 | 4 | 1008 | 128 | ||
| NM_022645.2 | NP_072171.1 | 3 | 470 | 128 | ||
| GenBank mRNA | BC022044.1 | - | 5 | 706 | 4769046 | 128 |
| BC035965.1 | - | 5 | 702 | 4731880 | 128 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

CSH2
Genomic Map