| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000880.hg.1 |
TC06000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000880.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000880.hg.1 |
ENST00000543875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000880.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000881.hg.1 |
TC06000083.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000881.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000881.hg.1 |
ENST00000543875.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06000881.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147617.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147617.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147617.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147617.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147617.hg.1 |
TC0600006950.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600147615.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147615.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147615.hg.1 |
TC0600006950.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600147616.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147616.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147616.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600147616.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600147616.hg.1 |
TC0600006950.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
ENST00000379426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600006950.hg.1 |
NM_001100829.2 |
Refseq |