Assay Details
| Gene Symbol: ZBTB16-RARA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_11_17_PLZF-RARa.0 | - | 198 | 135 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBTB16-RARA fusion| Gene Symbol: ZBTB16-RARA fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_11_17_PLZF-RARa.0 | - | 198 | 135 | ||
Set Membership: |
|