Assay Details
| Gene Symbol: MLL-MLLT3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_9_11_MLL-AF9_fusion_6A.0 | - | 99 | 103 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLL-MLLT3 fusion| Gene Symbol: MLL-MLLT3 fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_9_11_MLL-AF9_fusion_6A.0 | - | 99 | 103 | ||
Set Membership: |
|