Assay Details
| Gene Symbol: DEK-CAN fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_6_9_DEK-CAN_1.0 | - | 196 | 138 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEK-CAN fusion| Gene Symbol: DEK-CAN fusion | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | t_6_9_DEK-CAN_1.0 | - | 196 | 138 | ||
Set Membership: |
|