| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183009.hg.1 |
TC1200010822.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200183012.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183012.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183012.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200183012.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200183012.hg.1 |
TC1200010822.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000513533.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
ENST00000604081.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010822.hg.1 |
NR_047505.1 |
Refseq |