Assay Details
| Gene Symbol: HOTTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037843.3 | 1 - 2 | 101 | 71 | ||
| GenBank mRNA | AK093987.1 | - | 1 - 2 | 85 | 71 | |
| GU724873.1 | - | 1 - 2 | 101 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | homeobox A13 | HOXA13 |
HOX1, HOX1J |
-171.0 | Anti-sense | Chr. 7 - 27196880 - 27200106 on NCBI Build GRCh38 |
| Downstream | EVX1 antisense RNA | EVX1-AS | EVX1-AS1 | 38.6kb | Anti-sense | Chr. 7 - 27241429 - 27247229 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000170.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07002674.hg.1 | TC07000170.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002674.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002674.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002674.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002675.hg.1 | TC07000170.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002675.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002675.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002675.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07002675.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000616633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000616633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000618195.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000619957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000619957.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | ENST00000620415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148621.hg.1 | TC0700007007.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000421733.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000616633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000616633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000618195.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000619957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000619957.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | ENST00000620415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007007.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000620415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000472494.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000521028.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000605136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000614457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | ENST00000620415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | NR_037843.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700148618.hg.1 | TC0700007007.hg.1 | Clariom D TC |

HOTTIP
Genomic Map