Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HOXC cluster antisense RNA 2 | HOXC-AS2 | Sense | Chr. 12 - 53995208 - 53996785 on NCBI Build GRCh38 | ||
| Downstream | HOXC cluster antisense RNA 1 | HOXC-AS1 | 3.1kb | Sense | Chr. 12 - 53999022 - 54000010 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map