Assay Details
| Gene Symbol: TFAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001288.1 | 1 | 845 | 73 | ||
| GenBank mRNA | CV358951.1 | - | 1 | 469 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | PSMG3 antisense RNA 1 (head to head) | PSMG3-AS1 | -24.8kb | Sense | Chr. 7 - 1570073 - 1589626 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105375303 | LOC105375303 | 188.0 | Sense | Chr. 7 - 1616880 - 1631980 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000025.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 202236 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 253597 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 268878 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 2165 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3682 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3683 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P134727 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263472 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15900107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263464 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15900110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263458 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263463 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476240 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263474 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263460 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476201 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476203 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263481 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263472 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263485 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476218 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263482 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263468 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15900110 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476229 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263458 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263479 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476223 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15900107 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476231 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476238 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263464 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25476228 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05263453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263460 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263451 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15900126 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263501 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263506 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263485 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263478 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263479 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263509 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263486 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15900122 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263450 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263461 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263493 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15900125 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15900117 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15900120 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263484 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263456 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263475 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263483 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263511 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263448 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263513 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263453 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263491 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05263476 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P15964506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Genome U219 Array | 11763938_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11757245_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11757244_at | - | - |
| Affymetrix Human Genome U219 Array | 11757245_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11763938_at | - | - |
| Affymetrix Human Genome U219 Array | 11757244_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012255.hg.1 | ENST00000531132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000326.hg.1 | TC07000025.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07000326.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144768.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144768.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144768.hg.1 | TC0700006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144767.hg.1 | ENST00000412727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144767.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144767.hg.1 | ENST00000412727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700144767.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144767.hg.1 | TC0700006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700144766.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144766.hg.1 | NR_001288.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700144766.hg.1 | TC0700006526.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012255.hg.1 | ENST00000531132.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033867 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033866 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033866 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A033867 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3251695 | NR_001288.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3251695 | NR_001288.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3251695 | NR_001288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3251695 | NR_001288.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2138689 | NR_001288.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2138689 | NR_001288.1 | human_transcriptome |

TFAMP1
Genomic Map