Assay Details
| Gene Symbol: TERC | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001566.1 | LIT2117 telomerase RNA |
1 | 350 | 70 | ||
| GenBank mRNA | BC128029.1 | - | 1 | 350 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22897458 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22897453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02757652 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22897457 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22897452 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22897455 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3245248 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3701 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3700 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25297 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 19173 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3245248 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3245248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897512 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757696 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897497 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757708 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757705 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757691 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757694 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897515 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757695 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897500 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02757697 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22897496 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P820860 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757706 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757672 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757754 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757678 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757691 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757644 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757702 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15510027 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15510042 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757647 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757651 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757696 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757700 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757716 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757664 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757708 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757693 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757653 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757657 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757698 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757746 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757749 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757660 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15510041 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757674 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757684 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757676 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757740 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15510046 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757742 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757683 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02757704 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_001566_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8092001 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8092000 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC03001982.hg.1 | NR_001566.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC03001982.hg.1 | ENST00000363312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035489.hg.1 | TC03001982.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035489.hg.1 | ENST00000363312.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03035489.hg.1 | NR_001566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | ENST00000363312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | ENST00000602385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | NR_001566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | ENST00000363312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | ENST00000602385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | NR_001566.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300197774.hg.1 | TC0300013079.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013079.hg.1 | ENST00000363312.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013079.hg.1 | ENST00000602385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013079.hg.1 | ENST00000602385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013079.hg.1 | NR_001566.1 | Refseq |
| Human Genome U95 Set | 70723_at | - | - |
| Human Genome U95 Set | 70723_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021907 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A021907 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 4670343 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_24848 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1766573 | NR_001566.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766573 | BC128029.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1766573 | NR_001566.1 | human_transcriptome |

TERC
Genomic Map