Assay Details
| Gene Symbol: IFITM4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001590.1 | 1 | 54 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | HLA-F antisense RNA 1 | HLA-F-AS1 | -1.8kb | Sense | Chr. 6 - 29726601 - 29749049 on NCBI Build GRCh38 | |
| Downstream | HLA complex group 4 (non-protein coding) | HCG4 |
HCG4P10, HCGIV-10, HCGIV.9 |
39.9kb | Sense | Chr. 6 - 29791031 - 29793073 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P16124 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P16124 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 41743 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23699 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P16124 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P16124 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828562 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15798159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637790 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637757 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15798159 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637766 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637753 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828513 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637764 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637797 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637793 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637752 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828510 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828548 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828529 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637770 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637748 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828546 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828544 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828545 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828553 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637769 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828508 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828570 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828528 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637785 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637760 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637776 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828525 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828509 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637779 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04637767 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24828562 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P16124 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P16124 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637710 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637760 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637723 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637707 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637753 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637767 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637797 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637772 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15798100 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637715 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637782 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637783 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637711 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637705 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637730 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637718 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15798127 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637762 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637757 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637752 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637734 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637737 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637728 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637795 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637794 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637778 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637739 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637775 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637770 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637750 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637726 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637790 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637713 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637758 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637800 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15798163 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637780 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04637763 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029633 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2140341 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_12466 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1694058 | NR_001590.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1770071 | NR_001590.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1694058 | NR_001590.1 | human_transcriptome |

IFITM4P
Genomic Map