Assay Details
| Gene Symbol: DPRXP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002221.1 | 1 | 553 | 84 | ||
| GenBank mRNA | BQ448328.1 | - | 1 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 135 | RNF135 |
L13, MMFD, REUL, Riplet |
Sense | Chr. 17 - 30968642 - 30999911 on NCBI Build GRCh38 | |
| Downstream | neurofibromin 1 | NF1 |
NFNS, VRNF, WSS |
31.9kb | Sense | Chr. 17 - 31094927 - 31377677 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000359.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306048 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3736 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60177 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306048 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3306048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10281730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10281732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847321 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10281732 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30847315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10281730 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281720 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281719 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281727 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281732 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281731 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281729 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16968596 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281725 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281733 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16968599 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10281722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17004507.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004507.hg.1 | TC17000359.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700153580.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153580.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153580.hg.1 | TC1700007476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153582.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153582.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153582.hg.1 | TC1700007476.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700153581.hg.1 | ENST00000578053.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153581.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153581.hg.1 | ENST00000578053.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700153581.hg.1 | NR_002221.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700153581.hg.1 | TC1700007476.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009357 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A009357 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1690128 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_17928 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2051483 | BQ448328.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1739532 | NR_002221.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1739532 | BQ448328.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2051483 | NR_002221.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2051483 | BQ448328.1 | human_transcriptome |

DPRXP4
Genomic Map