Assay Details
| Gene Symbol: RFPL1S | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002727.2 | 1 | 2039 | 141 | |||
| GenBank mRNA | AJ010230.1 | - | LIT2045 RFPL1S |
1 | 2039 | 141 | |
| DA509757.1 | - | 1 | 419 | 141 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ret finger protein like 1 | RFPL1 | RNF78 | Anti-sense | Chr. 22 - 29403031 - 29442491 on NCBI Build GRCh38 | |
| Downstream | neurofilament heavy polypeptide | NEFH |
CMT2CC, NFH |
38.1kb | Anti-sense | Chr. 22 - 29480192 - 29491290 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000636.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22011065.hg.1 | ENST00000461286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011065.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011065.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011065.hg.1 | TC22000636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011066.hg.1 | ENST00000461286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011066.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011066.hg.1 | TC22000636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011064.hg.1 | ENST00000461286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011064.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22011064.hg.1 | TC22000636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160133.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160137.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160138.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000419368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008401.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160134.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160135.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | TC2200008401.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | NR_002727.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000461286.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | ENST00000539579.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200160136.hg.1 | NR_002727.2 | Refseq |

RFPL1S
Genomic Map