Assay Details
| Gene Symbol: CPS1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002763.2 | 1 | 447 | 137 | ||
| GenBank mRNA | AF090897.1 | - | 1 | 456 | 137 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | LanC like 1 | LANCL1 |
GPR69A, p40 |
-140.0kb | Anti-sense | Chr. 2 - 210431249 - 210477618 on NCBI Build GRCh38 |
| Downstream | erb-b2 receptor tyrosine kinase 4 | ERBB4 |
ALS19, HER4, p180erbB4 |
755.8kb | Anti-sense | Chr. 2 - 211375717 - 212539147 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001247.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF090897.1 transcripts.
Set Membership: |
|

CPS1-IT1
Genomic Map