Assay Details
| Gene Symbol: UBE2MP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002837.1 | 1 | 151 | 150 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | putative cyclin-Y-like protein 3 | LOC102724928 | -109.1kb | Anti-sense | Chr. 16 - 35030347 - 35060329 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 1566 | LINC01566 | 193.0kb | Anti-sense | Chr. 16 - 35363416 - 35391713 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16001077.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010133.hg.1 | ENST00000568522.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015150.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16015150.hg.1 | TC16001077.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000253023.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000593801.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000595957.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000596985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | ENST00000599829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011636.hg.1 | NM_003969.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178166.hg.1 | ENST00000568522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178166.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178166.hg.1 | ENST00000568522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600178166.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178166.hg.1 | TC1600010133.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010133.hg.1 | ENST00000568522.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010133.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178167.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178167.hg.1 | NR_002837.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600178167.hg.1 | TC1600010133.hg.1 | Clariom D TC |

UBE2MP1
Genomic Map