Assay Details
| Gene Symbol: KC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002838.1 | 4 - 5 | 640 | 125 | ||
| GenBank mRNA | AY762618.1 | - | 4 - 5 | 640 | 125 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1477 | LINC01477 | -1.4mb | Anti-sense | Chr. 18 - 40066286 - 40099233 on NCBI Build GRCh38 | |
| Downstream | phosphatidylinositol 3-kinase catalytic subunit type 3 | PIK3C3 |
VPS34, Vps34, hVps34 |
472.6kb | Anti-sense | Chr. 18 - 41955196 - 42084606 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000468.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18004804.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18004804.hg.1 | TC18000468.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | TC1800008536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158173.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000593577.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000595135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000596923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000599934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600183.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000600644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | ENST00000601523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008536.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | NR_002838.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | TC1800008536.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | ENST00000592302.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800158172.hg.1 | NR_002838.1 | Refseq |

KC6
Genomic Map