Assay Details
| Gene Symbol: SNORD103C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003066.1 | 1 | 31 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 103B |
SNORD103B | U103B | -19.0kb | Sense | Chr. 1 - 30949118 - 30949205 on NCBI Build GRCh38 |
| Downstream | Na+/K+ transporting ATPase interacting 1 | NKAIN1 | FAM77C | 211.5kb | Sense | Chr. 1 - 31179270 - 31239887 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002448.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4091 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139956 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139957 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139962 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139964 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139942 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139960 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139950 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139975 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139946 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139969 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139964 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139981 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139952 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139958 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139940 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139944 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139957 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139956 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139941 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045258 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139962 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139967 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117336 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139953 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139942 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139977 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139979 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045255 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139959 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117323 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045261 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117306 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117318 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117342 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117310 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117345 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117311 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117305 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117324 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117296 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15045257 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117347 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117329 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117315 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15045258 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01037668.hg.1 | TC01002448.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037668.hg.1 | ENST00000363311.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01037668.hg.1 | NR_003066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200603.hg.1 | NR_003066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200603.hg.1 | NR_003066.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100200603.hg.1 | TC0100013552.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3247303 | NR_003066.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3247303 | NR_003066.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3247303 | NR_003066.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3247303 | NR_003066.1 | human_transcriptome |

SNORD103C
Genomic Map